Learn More
Aviva Systems Biology Corp Ecsit Antibody - N-terminal region (ARP37175_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37175_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ecsit. It was validated on Western Blot. Ecsit,ECSIT homolog (Drosophila),Sitpec; Ecsit,evolutionarily conserved signaling intermediate in Toll pathway, mitochondrial,evolutionarily conserved signaling intermediate in Toll pathway, mitochondrial,signaling intermediate in Toll pathway-evolutionarily conservedThe immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: SitpecApplicationWB. Species reactivity: Mouse
Catalog No. 50-284-591