Learn More
Medchemexpress LLC Ecteinascidin 770 | 114899-80-8 | 99.8% | 770.85 | C40H42N4O10S | 50 MG
SDP
Catalog No. 5000212512
Encompass_Preferred
Ecteinascidin 770 is a tetrahydroisoquinoline alkaloid with potent cytotoxic and anticancer activity. It shows low-nanomolar IC50 values across multiple human cancer cell lines and is intended for preclinical in vitro research. The compound is provided as a high-purity solid or as a DMSO solution and is documented with CAS 114899-80-8, formula C40H42N4O10S, and molecular weight 770.85.
  • High purity (≈99.8%).
  • Potent cytotoxic activity with low-nanomolar IC50 in multiple human cancer cell lines.
  • Suitable for in vitro and preclinical research applications.
  • Available in mg-scale quantities and as DMSO solutions.
  • Characterized by CAS 114899-80-8, formula C40H42N4O10S, molecular weight 770.85.
Catalog No. 50-002-12512 Supplier Medchemexpress LLC Supplier No. HY10119150MG
Ecteinascidin 770 is a tetrahydroisoquinoline alkaloid with potent cytotoxic and anticancer activity. It shows low-nanomolar IC50 values across multiple human cancer cell lines and is intended for preclinical in vitro research. The compound is provided as a high-purity solid or as a DMSO solution and is documented with CAS 114899-80-8, formula C40H42N4O10S, and molecular weight 770.85.
  • High purity (≈99.8%).
  • Potent cytotoxic activity with low-nanomolar IC50 in multiple human cancer cell lines.
  • Suitable for in vitro and preclinical research applications.
  • Available in mg-scale quantities and as DMSO solutions.
  • Characterized by CAS 114899-80-8, formula C40H42N4O10S, molecular weight 770.85.