Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ecteinascidin 770 | 114899-80-8 | 99.8% | 770.85 g/mol | C40H42N4O10S | 1 ML
SDP

Catalog No. 5000211469
Encompass_Preferred

Ecteinascidin 770 (ET-770) is a tetrahydroisoquinoline alkaloid used in preclinical anticancer research. It exhibits potent cell-based activity, inhibiting U373MG cells with a reported IC50 of 4.83 nM. The material is supplied as high-purity solid and as DMSO solutions for biochemical and cellular studies.

  • Potent anticancer activity, reported IC50 4.83 nM against U373MG cells.
  • High purity (~99.8%) suitable for research applications.
  • Available as 10 mM solution in DMSO or as solid quantities.
  • Molecular weight 770.85 g/mol; formula C40H42N4O10S.
  • Intended for in vitro research use only.

Catalog No. 50-002-11469 Supplier Medchemexpress LLC Supplier No. HY10119110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ecteinascidin 770 (ET-770) is a tetrahydroisoquinoline alkaloid used in preclinical anticancer research. It exhibits potent cell-based activity, inhibiting U373MG cells with a reported IC50 of 4.83 nM. The material is supplied as high-purity solid and as DMSO solutions for biochemical and cellular studies.

  • Potent anticancer activity, reported IC50 4.83 nM against U373MG cells.
  • High purity (~99.8%) suitable for research applications.
  • Available as 10 mM solution in DMSO or as solid quantities.
  • Molecular weight 770.85 g/mol; formula C40H42N4O10S.
  • Intended for in vitro research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.