Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ecteinascidin 770 | 114899-80-8 | 99.8% | 770.85 g/mol | C40H42N4O10S | 10 MG
SDP

Catalog No. 5000213473
Encompass_Preferred

Ecteinascidin 770 is a 1,2,3,4-tetrahydroisoquinoline alkaloid natural product used in preclinical cancer research for its potent cytotoxic and apoptosis-inducing activities. It is typically handled as a solid and prepared in solvent for in vitro assays, showing low-nanomolar inhibitory potency across several human tumor cell lines.

  • Induces apoptosis in tumor cell lines.
  • Exhibits low-nanomolar potency (reported IC50 values in multiple lines).
  • Molecular formula C40H42N4O10S and molecular weight ~770.85 g/mol.
  • Soluble in DMSO for biological testing.
  • Supplied as a small solid vial suitable for research use.

Catalog No. 50-002-13473 Supplier Medchemexpress LLC Supplier No. HY10119110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ecteinascidin 770 is a 1,2,3,4-tetrahydroisoquinoline alkaloid natural product used in preclinical cancer research for its potent cytotoxic and apoptosis-inducing activities. It is typically handled as a solid and prepared in solvent for in vitro assays, showing low-nanomolar inhibitory potency across several human tumor cell lines.

  • Induces apoptosis in tumor cell lines.
  • Exhibits low-nanomolar potency (reported IC50 values in multiple lines).
  • Molecular formula C40H42N4O10S and molecular weight ~770.85 g/mol.
  • Soluble in DMSO for biological testing.
  • Supplied as a small solid vial suitable for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.