Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ecteinascidin 770 (ecteinascidine 770) | 114899-80-8 | 99.8% | 770.85 g·mol⁻¹ | C40H42N4O10S | 1 MG
SDP

Catalog No. 5000212843
Encompass_Preferred

Ecteinascidin 770 (ET-770) is a 1,2,3,4-tetrahydroisoquinoline alkaloid with potent cytotoxic activity used in preclinical cancer research. It exhibits low-nanomolar IC50 values against multiple human cell lines and is supplied as a high-purity solid for laboratory experiments.

  • 1 mg solid, white to yellow.
  • Molecular weight 770.85 g·mol⁻¹.
  • Chemical formula C40H42N4O10S.
  • Purity approximately 99.8%.
  • Store at -20°C; protect from light and store under inert gas; in solvent: -80°C (up to 6 months), -20°C (up to 1 month).
  • Demonstrates potent cytotoxicity; inhibits U373MG cells with IC50 = 4.83 nM.
  • Datasheet and safety data sheet available for handling and hazard information.

Catalog No. 50-002-12843 Supplier Medchemexpress LLC Supplier No. HY1011911MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ecteinascidin 770 (ET-770) is a 1,2,3,4-tetrahydroisoquinoline alkaloid with potent cytotoxic activity used in preclinical cancer research. It exhibits low-nanomolar IC50 values against multiple human cell lines and is supplied as a high-purity solid for laboratory experiments.

  • 1 mg solid, white to yellow.
  • Molecular weight 770.85 g·mol⁻¹.
  • Chemical formula C40H42N4O10S.
  • Purity approximately 99.8%.
  • Store at -20°C; protect from light and store under inert gas; in solvent: -80°C (up to 6 months), -20°C (up to 1 month).
  • Demonstrates potent cytotoxicity; inhibits U373MG cells with IC50 = 4.83 nM.
  • Datasheet and safety data sheet available for handling and hazard information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.