Learn More
Cayman Chemical Ecto5nucleotIdaseCD73 Rab
SDP
Catalog No. 502913345
Encompass_Preferred
A carbamate pesticide inhibits mycelial growth of the plant pathogenic fungus A rabiei on potato dextrose agar (EC50 2 19 Ug/ml) decreases the fecundity of red ant (M rubra) queens at 1 mg/ml in the drinking water induces the production of ROS apoptosis and DNA damage in H9c2 rat cardiomyocytes
Catalog No. 50-291-3345 Supplier Cayman Chemical Supplier No. 35613100 UL
A carbamate pesticide inhibits mycelial growth of the plant pathogenic fungus A rabiei on potato dextrose agar (EC50 2 19 Ug/ml) decreases the fecundity of red ant (M rubra) queens at 1 mg/ml in the drinking water induces the production of ROS apoptosis and DNA damage in H9c2 rat cardiomyocytes