Learn More
Apexbio Technology LLC ECTOINE-200MG
SDP
Catalog No. 5000569321
Encompass_Preferred
Ectoine (CAS No 96702-03-3) is a prominent molecule in the field of biomedical research identified as a natural compound derived from extremophilic microorganisms It functions primarily as a protective osmolyte stabilizing proteins and cellular structures under stress conditions by influencing hydration shells Ectoine demonstrates significant in vitro activity within the nanomolar to low micromolar range showcasing its efficacy in various experimental settings It is widely utilized in cell models to investigate its protective effects against environmental stressors and its applications extend to animal studies and drug screening processes With its ability to mitigate cellular stress Ectoine serves as a potential therapeutic agent in studies focused on inflammation oxidative stress and skin barrier repair Experimental concentrations typically depend on the specific research design providing flexibility in its application across diverse biomedical research contexts
Catalog No. 50-005-69321 Supplier Apexbio Technology LLC Supplier No. C7041200
Ectoine (CAS No 96702-03-3) is a prominent molecule in the field of biomedical research identified as a natural compound derived from extremophilic microorganisms It functions primarily as a protective osmolyte stabilizing proteins and cellular structures under stress conditions by influencing hydration shells Ectoine demonstrates significant in vitro activity within the nanomolar to low micromolar range showcasing its efficacy in various experimental settings It is widely utilized in cell models to investigate its protective effects against environmental stressors and its applications extend to animal studies and drug screening processes With its ability to mitigate cellular stress Ectoine serves as a potential therapeutic agent in studies focused on inflammation oxidative stress and skin barrier repair Experimental concentrations typically depend on the specific research design providing flexibility in its application across diverse biomedical research contexts