Learn More
Perkin Elmer US LLC Twist Type Torch Injector Support Adapter for ELAN 9000/6x00/5000/DRC-e
Catalog No. 509045544
Encompass_Preferred
This Injector Support Adapter is for use with ELAN 9000/6x00/5000/DRC-e ICP-MS series instruments. The Twist Type Torch Adapter does not include O-rings.
Catalog No. 50-904-5544 Supplier Perkin Elmer US LLC Supplier No. N8122007
This Injector Support Adapter is for use with ELAN 9000/6x00/5000/DRC-e ICP-MS series instruments. The Twist Type Torch Adapter does not include O-rings.