Learn More
Medchemexpress LLC Eculizumab 10mg | 10mg
SDP
Catalog No. 5000323369
Encompass_Preferred
Eculizumab (Anti-Human C5 Humanized Antibody) is a long-acting humanized monoclonal antibody targeted against complement C5 Eculizumab inhibits the cleavage of C5 into C5a and C5b and inhibits deployment of the terminal complement system including the formation of membrane attack complex (MAC) Eculizumab prevents anti-ganglioside antibody-mediated neuropathy in mice Eculizumab can be used in hemolysis studies[1][2]
Catalog No. 50-003-23369 Supplier Medchemexpress LLC Supplier No. HYP991410MG
Eculizumab (Anti-Human C5 Humanized Antibody) is a long-acting humanized monoclonal antibody targeted against complement C5 Eculizumab inhibits the cleavage of C5 into C5a and C5b and inhibits deployment of the terminal complement system including the formation of membrane attack complex (MAC) Eculizumab prevents anti-ganglioside antibody-mediated neuropathy in mice Eculizumab can be used in hemolysis studies[1][2]