Learn More
Labtech Inc ED36S Digiblock Digester, 36 positions for 50ml digestion tubes, Max Temperature: 210℃, Temp accuracy: ±0.2℃, Heating power: 1400W, Heating material: PTFE coating/Al, Power Supply: 230V, 50/60Hz
Catalog No. 5000496178
Encompass_Preferred
ED36S Digiblock Digester, 36 positions for 50ml digestion tubes, Max Temperature: 210℃, Temp accuracy: ±0.2℃, Heating power: 1400W, Heating material: PTFE coating/Al, Power Supply: 230V, 50/60Hz
Catalog No. 50-004-96178 Supplier Labtech Inc Supplier No. LH7016
ED36S Digiblock Digester, 36 positions for 50ml digestion tubes, Max Temperature: 210℃, Temp accuracy: ±0.2℃, Heating power: 1400W, Heating material: PTFE coating/Al, Power Supply: 230V, 50/60Hz