Learn More
Labtech Inc ED54S Digiblock Digester, 54 positions for 50ml digestion tubes, Max Temperature: 240℃, Temp accuracy: ±0.2℃, Heating power: 3000W, Heating material: graphite PTFE coating/Al, Power Supply: 230V, 50/60Hz
Catalog No. 5000496194
Encompass_Preferred
ED54S Digiblock Digester, 54 positions for 50ml digestion tubes, Max Temperature: 240℃, Temp accuracy: ±0.2℃, Heating power: 3000W, Heating material: graphite PTFE coating/Al, Power Supply: 230V, 50/60Hz
Catalog No. 50-004-96194 Supplier Labtech Inc Supplier No. LH7018
ED54S Digiblock Digester, 54 positions for 50ml digestion tubes, Max Temperature: 240℃, Temp accuracy: ±0.2℃, Heating power: 3000W, Heating material: graphite PTFE coating/Al, Power Supply: 230V, 50/60Hz