Learn More
Enzo Life Sciences EDA-A1 (soluble) (human), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX522038C010
Encompass_Preferred
Alternative name: Ectodysplasin A1. Concentration: 0.1mg/ml after reconstitution. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in E. coli. The extracellular domain of human EDA-A1 (ectodysplasin-A1) (aa 245-391) is fused at the N-terminus to a linker peptide (9 aa) and a FLAG-tag. Specificity: Binds to human EDAR (ectodysplasin-A1 receptor). Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-200-9384