Learn More
Aviva Systems Biology Corp EDA Antibody - middle region (ARP44519_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44519_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EDA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EDA. Alternate names: ED1, ED1-A1, ED1-A2, EDA1, EDA2, HED, XHED, XLHED, ODT1, STHAGX1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58833