Learn More
Medchemexpress LLC Eda-da tfa | 87156-01-2 | 95.0% | 298.22 | 5 MG
SDP
Catalog No. 5000359917
Encompass_Preferred
EDA-DA is a N-terminally tagged dipeptide probe used to label Peptidoglycan (PG) of bacteria. It functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • N-terminally tagged dipeptide probe
  • Labels peptidoglycan of bacteria
  • Click chemistry reagent with an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-59917 Supplier Medchemexpress LLC Supplier No. HY147097A5MG
EDA-DA is a N-terminally tagged dipeptide probe used to label Peptidoglycan (PG) of bacteria. It functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • N-terminally tagged dipeptide probe
  • Labels peptidoglycan of bacteria
  • Click chemistry reagent with an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)