Learn More
Cayman Chemical Edaglitazone, Purity: ≥98%, CAS: 213411-83-7, 10 mg
SDP
Catalog No. 502983345
Encompass_Preferred
A PPARγ agonist; activates PPARγ in a reporter assay using CV-1 cells expressing murine PPARγ2 from 0.1-1 μM; increases [3H]2-deoxyglucose transport in the presence and absence of insulin and increases glycogen synthesis in the presence of insulin in soleus muscle strips isolated from obese rats at 1, 5, and 25 μM
Catalog No. 50-298-3345 Supplier Cayman Chemical Supplier No. 1911710 MG
A PPARγ agonist; activates PPARγ in a reporter assay using CV-1 cells expressing murine PPARγ2 from 0.1-1 μM; increases [3H]2-deoxyglucose transport in the presence and absence of insulin and increases glycogen synthesis in the presence of insulin in soleus muscle strips isolated from obese rats at 1, 5, and 25 μM