Learn More
Medchemexpress LLC HY-D1080 100mg , EDANS CAS:50402-56-7 Purity:>98%
SDP
Catalog No. 502261542
Encompass_Preferred
HY-D1080 100mg EDANS CAS: 50402-56-7 EDANS (1,5-EDANS) is a novel and quenched fluorogenic substrate for assaying retroviral protease by resonance energy transfer (RET). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1542 Supplier Medchemexpress LLC Supplier No. HYD1080100MG
HY-D1080 100mg EDANS CAS: 50402-56-7 EDANS (1,5-EDANS) is a novel and quenched fluorogenic substrate for assaying retroviral protease by resonance energy transfer (RET). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.