Learn More
Aat Bioquest EDANS sodium salt [5-((2-Aminoethyl)aminonaphthalene-1-sulfonic acid, sodium salt] *CAS 100900-07-0*
SDP
Catalog No. 501952937
Encompass_Preferred
EDANS sodium salt is much more water-soluble than EDANS acid. It is one of the most popular donors for developing FRET-based nucleic acid probes (such as Molecular Beacons) and protease substrates.
Catalog No. 50-195-2937 Supplier Aat Bioquest Supplier No. 615
EDANS sodium salt is much more water-soluble than EDANS acid. It is one of the most popular donors for developing FRET-based nucleic acid probes (such as Molecular Beacons) and protease substrates.