Learn More
Apexbio Technology LLC Edaravone 89-25-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A50595.1
Encompass_Preferred
Edaravone (CAS 89-25-8) is a small-molecule antioxidant and free radical scavenger that mitigates cell injury primarily by intercepting reactive oxygen species and suppressing oxidative stress responses Its cytoprotective mechanism has been linked to the inhibition of lipid peroxidation pathways potentially involving suppression of lipoxygenase activity Edaravone has been investigated for neuroprotective effects notably demonstrating protection against neurotoxicity induced by MPTP a well-established model to study Parkinson s disease pathology This compound serves as a significant research tool for examining oxidative injury in ischemic and neurodegenerative conditions
Catalog No. 50-000-62497