Learn More
Medchemexpress LLC HY-B0099 5g Medchemexpress, Edaravone CAS:89-25-8 Purity:>98%
SDP
Catalog No. 501932899
Encompass_Preferred
Medchemexpress, HY-B0099 5g Edaravone CAS:89-25-8 Edaravone is a strong novel free radical scavenger, and inhibits MMP -9-related brain hemorrhage in rats treated with tissue plasminogen activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2899 Supplier Medchemexpress LLC Supplier No. HYB00995G
Medchemexpress, HY-B0099 5g Edaravone CAS:89-25-8 Edaravone is a strong novel free radical scavenger, and inhibits MMP -9-related brain hemorrhage in rats treated with tissue plasminogen activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.