Learn More
Medchemexpress LLC Edaravone | 89-25-8 | MFCD00003138 | 99.9% | C10H10N2O | 25 G
SDP
Catalog No. 5000359015
Encompass_Preferred
Edaravone is a potent free radical scavenger and neuroprotective agent. It exhibits preventative and therapeutic effects against glutamate toxicity by reducing apoptosis and necrosis. It also inhibits MMP-9-related brain hemorrhage and ameliorates neuronal damage in brain ischemia. This compound is intended for research use only, supporting studies on neuroprotection and free radical scavenging.
  • Strong free radical scavenger
  • Inhibits MMP-9-related brain hemorrhage
  • Demonstrates neuroprotective activity against H2O2-induced cell damage
  • Shows neuroprotective activity against glutamate-induced toxicity
  • Inhibits arachidonic acid and adenosine diphosphate-induced platelet aggregation
  • Reduces astrogliosis and apoptosis
  • Increases protein expression of TGF-β1, VEGF, and MMP9
  • Suppresses TNF-α expression
Catalog No. 50-003-59015 Supplier Medchemexpress LLC Supplier No. 25GHYB0099
Edaravone is a potent free radical scavenger and neuroprotective agent. It exhibits preventative and therapeutic effects against glutamate toxicity by reducing apoptosis and necrosis. It also inhibits MMP-9-related brain hemorrhage and ameliorates neuronal damage in brain ischemia. This compound is intended for research use only, supporting studies on neuroprotection and free radical scavenging.
  • Strong free radical scavenger
  • Inhibits MMP-9-related brain hemorrhage
  • Demonstrates neuroprotective activity against H2O2-induced cell damage
  • Shows neuroprotective activity against glutamate-induced toxicity
  • Inhibits arachidonic acid and adenosine diphosphate-induced platelet aggregation
  • Reduces astrogliosis and apoptosis
  • Increases protein expression of TGF-β1, VEGF, and MMP9
  • Suppresses TNF-α expression