Learn More
Medchemexpress LLC Edaravone-d5 | 1228765-67-0 | 99.5% | C10H5D5N2O | 1 MG
SDP
Catalog No. 5000433511
Encompass_Preferred
Edaravone-d5 is a deuterium-labeled form of Edaravone, a novel and strong free radical scavenger. It effectively inhibits MMP-9-related brain hemorrhage, particularly in rats treated with tissue plasminogen activator.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-33511 Supplier Medchemexpress LLC Supplier No. HYB0099S1MG
Edaravone-d5 is a deuterium-labeled form of Edaravone, a novel and strong free radical scavenger. It effectively inhibits MMP-9-related brain hemorrhage, particularly in rats treated with tissue plasminogen activator.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS