Learn More
Abcam Edaravone (MCI-186), antio x idant, 250MG
Catalog No. 502707396
Encompass_Preferred
MW 174.2 Da, Purity >99%. Free radical scavenger and potent antioxidant. Inhibits production of reactive oxygen species, apoptosis and the lipoxygenase system.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7396 Supplier Abcam Supplier No. AB120645250MG
MW 174.2 Da, Purity >99%. Free radical scavenger and potent antioxidant. Inhibits production of reactive oxygen species, apoptosis and the lipoxygenase system.
The product is subject to the following: Abcam Restricted Use Statement