Learn More
Bioworld EDC (1-ethyl-3-(3-dimethylaminopropyl) Carbodiimide Hydrochloride), 5 g
Supplier:  Bioworld 405000041
Encompass_Preferred
An enzyme inhibitor EDC is a water soluble derivative of carbodiimide. Carbodimides couple carboxyl to primary amines resulting in the formation of amide bonds Carbodimides are unlike other conjugation reactions in that no spacer arms exists between the molecules being coupled.Appearance: White Crystalline PowderBoiling Point : 197.7 °CDensity : 0.88 g/cm3HS Code : 29252000IUPAC_Name: N-[3-(dimethylamino)propyl]-N’-ethylcarbodiimide hydrochlorideMelting Point: 110-115 °CRTECS Number: FF2200000
Catalog No. 50-255-936