Learn More
Slice™ EDC Folding Knife
Catalog No. 18222854
Change view
Click to view available options
Blade Material:
Zirconium Oxide
Catalog No. Blade Material
18-222-854 Zirconium Oxide
1 options
Catalog No. 18-222-854 Supplier Slice™ Supplier No. 10495
Add to Cart
Add to Cart
The Slice™ 10495 EDC Folding Knife is a stylish, modernized version of the popular folding knife that exemplifies craftsmanship and innovative design.
  • Installed blade type: Rounded tip No. 10404
  • Sturdy metal handle with locking liner
  • Spring-assist deployment
  • Ambidextrous design
  • Lanyard hole
  • Finger-friendly™ blade cuts effectively
  • Reduces injuries, lowers costs
  • Non-sparking, non-conductive, non-magnetic blade
  • Chemically inert blade never rusts
  • Oil- and lubricant-free blade
  • 100 percent zirconium oxide blade
  • Blade edge lasts up to 11.2x longer than metal
  • Fewer blade changes = fewer injuries
  • No-tool blade change
Specifications
Product Type Folding Knife
Blade Material Zirconium Oxide
For Use With (Equipment) Compatible blades: 10404, 10408
Handle Material Steel, Aluminum, NGF, TPFE, Nylon
Dimensions (L x W x H) 100.0 x 26.0 x 12.0 mm
Weight (Metric) 53.4 g