Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLD PHARMATECH CO LIMITED EDC·xHCl, 7084-11-9, Purity: 99%, 100g

Catalog No. NC3968952
Encompass_Preferred

EDC·xHCl, 7084-11-9, Purity: 99%, 100g

Catalog No. NC3968952 Supplier BLD PHARMATECH CO LIMITED Supplier No. BD00769847100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EDC·xHCl, 7084-11-9, Purity: 99%, 100g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.