Learn More
Aviva Systems Biology Corp EDEM1 Antibody - N-terminal region (ARP47054_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47054_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EDEM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EDEM1. Alternate names: EDEM, KIAA0212ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-59460