Learn More
Aviva Systems Biology Corp EDEM3 Antibody - C-terminal region (ARP62508_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62508_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EDEM3. It was validated on Western Blot. . Alternate names: C1orf22ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-287-700