Learn More
U.S. Pharmacopeia Edetate Disodium, 6381-92-6, MFCD00150037, 200mg
Catalog No. NC9116103
Encompass
Molecular formula C10H16N2O8.2H2O.2Na, Molecular weight 372.24, Melting Point 485.6 °F (252 °C) (decomposes), Synonyms: Disodium EDTA dihydrate
Catalog No. NC9116103 Supplier U.S. Pharmacopeia Supplier No. 1233009
Molecular formula C10H16N2O8.2H2O.2Na, Molecular weight 372.24, Melting Point 485.6 °F (252 °C) (decomposes), Synonyms: Disodium EDTA dihydrate