Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edetate Disodium U.S.P. GenAR, Macron Fine Chemicals™

Catalog No. MK7727500 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK-7727-500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-7727-500 Supplier Avantor Supplier No. 772704
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 6381-92-6
Packaging POLYSTORMOR
Molecular Formula C10H18N2Na2O10
Quantity 500 g
MDL Number MFCD00150037,MFCD00003541
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Molecular Weight (g/mol) 372.24
PubChem CID 44120005
Formula Weight 372.24
Percent Purity ≥99.0%
Grade USP
Physical Form Crystalline Powder
Chemical Name or Material Edetate Disodium
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.