Learn More
Aviva Systems Biology Corp EDF1 Antibody - N-terminal region (ARP35695_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35695_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EDF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EDF1. Alternate names: EDF-1, MBF1, MGC9058ApplicationIF, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-55820