Learn More
Aviva Systems Biology Corp EDG8 Antibody - N-terminal region (ARP42662_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42662_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EDG8. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human EDG8. Alternate names: Edg-8, S1P5, S1PR5, SPPR-1, SPPR-2, EDG8ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-58296