Learn More
Biocare Medical, LLC Eosin Y - 1 Gallon
SDP
Catalog No. 5082884
Encompass_Preferred
This modified alcoholic Eosin Y includes the addition of stabilizers for a prolonged shelf life. It is intended for use in the histologic demonstration of cytoplasm. Erythrocytes, collagen and the cytoplasm of muscle or epithelial cells should stain with three different intensities of pink.
Catalog No. 50-828-84 Supplier Biocare Medical, LLC Supplier No. HTEGL
This modified alcoholic Eosin Y includes the addition of stabilizers for a prolonged shelf life. It is intended for use in the histologic demonstration of cytoplasm. Erythrocytes, collagen and the cytoplasm of muscle or epithelial cells should stain with three different intensities of pink.