Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Water

Catalog No. EBT0779
Change view
Click to view available options
Water
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EB-T0779 Water
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-T0779 Supplier Edge Biologicals Supplier No. T0779
This item is not returnable. View return policy

Water for solution dilutions

 

  • Used in making dilutions of samples in preparation for microbial testing of the sample (can also be used for diluting sterile reagents and microorganism preparations)
  • Water does not contain Sodium Chloride
  • Autoclavable
  • Comes in screw-capped glass test tubes, 16 × 125 mm (0.630 × 4.92 in.)

 

Specifications

Content And Storage 8-25°C
Form Liquid
For Use With (Application) Diluting sterile reagents and microorganisms preparations
Packaging Type Tube
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.