Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals 20mL Deep Trypticase Soy Agar

Catalog No. EBT0753
Change view
Click to view available options
Quantity:
20 mL deep
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EB-T0753 20 mL deep
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EB-T0753 Supplier Edge Biologicals Supplier No. T0753
Add to Cart
Add to Cart

For isolation, cultivation and storage (in slant form) of fastidious and non-fastidious microorganisms

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Media Type Trypticase Soy Agar
For Use With (Application) Isolation, cultivation and storage (in slant form) of fastidious and non-fastidious microorganisms
No. per Pack 10
Quantity 20 mL deep
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.