Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Bile Esculin Agar Slant

Catalog No. EBT0081
Change view
Click to view available options
Color:
Bright Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
EB-T0081 Bright Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EB-T0081 Supplier Edge Biologicals Supplier No. T0081

General purpose medium for the cultivation of a wide variety of bacteria, yeast and fungi

  • Prepared suspension
  • Color: Dark amber
  • Sterile
  • Autoclavable
  • Final pH is 6.83
  • Pancreatic digest of Gelatin 5g/L
  • Beef extract 3g/L
  • Oxgall 20g/L
  • Ferric Citrate 0.5g/L
  • Esculin 1g/L
  • Agar 14g/L
  • Deionized Water 1000mL

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Color Bright Red
Content And Storage 8°C to 12°C
Form Suspension
Manufacturing Origin United States
Media Type Bile Esculin Agar
Product Type Agar
For Use With (Application) Isolation and presumptive identification of group D streptococci
Packaging Type Tube
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.