Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Brain Heart Infusion Agar Slant

Catalog No. EBT0006
Change view
Click to view available options
Color:
Light Amber
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
EB-T0006 Light Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EB-T0006 Supplier Edge Biologicals Supplier No. T0006
Add to Cart
Add to Cart

General purpose medium used for the cultivation of a wide variety of bacteria, yeast and fungi

  • Color: Light amber
  • Agar suspension soluble in water
  • Sterile, autoclavable
  • Final pH is 7.46

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Color Light Amber
Content And Storage 8°C to 12°C
Detects Bacteria, Yeast and Fungi
Form Suspension
Format Slant
Manufacturing Origin United States
Media Type BHI Agar
Product Type Agar Slants
Test Type Microbial Cultivation
Formulation Pork Brains infusion from 200 g 7.7g/L, Pork Peptone No. 2 10g/L, Sodium Phosphate 2.5g/L, Disodium Phosphate 2.5g/L, Pork Brains infusion from 250 g 9.8g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Agar 15g/L, Deionized Water 1000mL
For Use With (Application) Used for culture any of a variety of anaerobes
Includes 10 Tubes/Pk.
No. per Pack 10
Packaging Type Tube
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.