Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Brain Heart Infusion Broth

Catalog No. EBT0041
Click to view available options
Quantity:
0.5 mL
10 mL
Color:
Amber
Light Amber

Catalog No. EB-T0041

Supplier: Edge Biologicals T0041

Only null left
Add to Cart
Add to Cart

General purpose medium for the cultivation of a variety of bacteria, yeast and fungi

  • Sterile
  • Autoclavable
  • Color: Clear amber
  • Brain heart infusion from (solids) 6g/L
  • Peptic digest of Animal Tissue 6g/L
  • Pancreatic digest of Gelatin 14.5g/L
  • Sodium Chloride 5g/L
  • Dextrose 3g/L
  • Disodium Phosphate 2.5g/L
  • Deionized Water 1000mL

0.5mL

  • Final pH is 7.24
10mL
    • Final pH is 7.39

    Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance M22-A3
Color Amber
Content And Storage 8°C to 12°C
Form Liquid
Manufacturing Origin United States
Media Type BHI Broth
For Use With (Application) Cultivation of variety of bacteria, Yeast and fungi including streptococci and meningococci, antimicrobial susceptibility testing and Blood culture
Quantity 0.5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.