Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Brain Heart Infusion Broth

Catalog No. EBT0061
Change view
Click to view available options
Quantity:
0.5 mL
10 mL
Color:
Amber
Light Amber
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color
EB-T0061 10 mL Light Amber
EB-T0041 0.5 mL Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. EB-T0061 Supplier Edge Biologicals Supplier No. T0061
Add to Cart
Add to Cart
This item is not returnable. View return policy

General purpose medium for the cultivation of a variety of bacteria, yeast and fungi

  • Sterile
  • Autoclavable
  • Color: Clear amber
  • Brain heart infusion from (solids) 6g/L
  • Peptic digest of Animal Tissue 6g/L
  • Pancreatic digest of Gelatin 14.5g/L
  • Sodium Chloride 5g/L
  • Dextrose 3g/L
  • Disodium Phosphate 2.5g/L
  • Deionized Water 1000mL

0.5mL

  • Final pH is 7.24
10mL
    • Final pH is 7.39

    Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance M22-A3
Color Light Amber
Content And Storage 8°C to 12°C
Form Liquid
Manufacturing Origin United States
Media Type BHI Broth
For Use With (Application) Cultivation of variety of bacteria, Yeast and fungi including streptococci and meningococci, antimicrobial susceptibility testing and Blood culture
No. per Pack 10
Packaging Type Tube, Glass
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.