Learn More
Edge Biologicals Brain Heart Infusion Broth
Description
- Sterile
- Autoclavable
- Color: Clear amber
- Brain heart infusion from (solids) 6g/L
- Peptic digest of Animal Tissue 6g/L
- Pancreatic digest of Gelatin 14.5g/L
- Sodium Chloride 5g/L
- Dextrose 3g/L
- Disodium Phosphate 2.5g/L
- Deionized Water 1000mL
0.5mL
- Final pH is 7.24
- Final pH is 7.39
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Specifications
| Certifications/Compliance | M22-A3 |
| Color | Amber |
| Content And Storage | 8°C to 12°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | BHI Broth |
| For Use With (Application) | Cultivation of variety of bacteria, Yeast and fungi including streptococci and meningococci, antimicrobial susceptibility testing and Blood culture |
| Quantity | 0.5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.