Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Campy Blood Agar Plate

Catalog No. EBP4120
Change view
Click to view available options
Campy Blood Agar Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EB-P4120 Campy Blood Agar Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-P4120 Supplier Edge Biologicals Supplier No. P4120
This item is not returnable. View return policy

Used in the selective isolation and cultivation of Campylobacter species from human fecal specimens

  • Colors: Bright red, opaque
  • Sterile
  • Autoclavable
  • Format: Suspension
  • Final pH: 7.00
  • Pancreatic digest of Casein 10g/L
  • Peptic digest of Animal tissue 10g/L
  • Sodium Chloride 5g/L, Dextrose 1g/L
  • Yeast extract 2g/L, Sodium Bisulfite 0.1g/L
  • Amphotericin B 4mg/L, Cephalothin 15mg/L
  • Trimethoprim 5mg/L, Vancomycin 10mg/L
  • Polymyxin B 2500U

Specifications

Certifications/Compliance Certificate of analysis
Color Red
Content And Storage 8°C to 12°C
Form Liquid
Manufacturing Origin United States
Media Type Campylobacter Blood Agar
Sterility Sterile
For Use With (Application) Cultivation of Campylobacter
Packaging Type Plate
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.