Learn More
Edge Biologicals Cetrimide Agar
Catalog No. EBP4275
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
EB-P4275 10/Pk.
1 options
Catalog No. EB-P4275 Supplier Edge Biologicals Supplier No. P4275
Add to Cart
Add to Cart
For selective isolation and presumptive identification of Pseudomonas aeruginosa
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Quantity 10/Pk.