Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Chocolate Agar Plate

Catalog No. EBP3125
Change view
Click to view available options
Certifications/Compliance:
Certificate of analysis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
EB-P3125 Certificate of analysis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EB-P3125 Supplier Edge Biologicals Supplier No. P3125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used in qualitative procedures for the isolation and cultivation of fastidious organisms

  • Prepared agar plate
  • For isolation/cultivation of organisms such as pathogenic Neisseria and Haemophilus from clinical and nonclinical specimens
  • Color: Opaque brown
  • Sterile
  • Autoclavable
  • Final pH: 7.20
  • Proteose Peptone #315g/L
  • Corn Starch 1g/L
  • Potassium Phosphate, dibasic 4g/L
  • Potassium Phosphate, monobasic 1g/L
  • Sodium Chloride 5g/L
  • Agar 10g/L
  • Hemoglobin 10g/L
  • Deionized Water 1000mL
  • Supplement 10mL, Supplement ingredients/liter,
  • Adenine Sulfate

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Color Red
Content And Storage 8°C to 12°C
Country of Origin United States
Detects Fastidious Organisms, Neisseria, Haemophilus
Form Liquid
Format Plate
Manufacturing Origin United States
Media Type Chocolate Agar
Product Type Agar Plate
Sterility Sterile
Target Organism Class Fastidious Organisms
For Use With (Application) Growth of Neiseria and Hemophilus species
Packaging Type Plate
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.