Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Chocolate Agar Slant

Catalog No. EBT0120
Change view
Click to view available options
Color:
Brown
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
EB-T0120 Brown
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EB-T0120 Supplier Edge Biologicals Supplier No. T0120
Add to Cart
Add to Cart

For the isolation and cultivation of fastidious organisms

  • Prepared agar slant
  • For organisms such as pathogenic italics (begin)Neisseriaitalics (end) and italics (begin)Haemophilusitalics (end) from clinical and nonclinical specimens
  • Color: Opaque brown
  • Proteose Peptone #3 15g/L
  • Corn Starch 1g/L
  • Potassium Phosphate 4g/L dibasic,
  • Potassium Phosphate 1g/L monobasic,
  • Sodium Chloride 5g/L Agar 10g/L
  • Hemoglobin 10g/L
  • Deionized Water 1000mL,
  • Supplement 10mL,
  • Supplement ingredients: Adenine Sulfate 1g/L p-Ami
  • Sterile
  • Autoclavable
  • Final pH is 7.15

Proteose Peptone #3 15g/L Corn Starch 1g/L Potassium Phosphate 4g/L dibasic, Potassium Phosphate 1g/L monobasic, Sodium Chloride 5g/L Agar 10g/L Hemoglobin 10g/L Deionized Water 1000mL, supplement 10mL, supplement ingredients/L, Adenine Sulfate 1g/L p-Ami

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Appearance Brown Agar Slant
Color Brown
Content And Storage 8°C to 12°C
Detects Fastidious Organisms, Neisseria, Haemophilus
Form Suspension
Format Agar Slant
Manufacturing Origin United States
Media Type Chocolate Agar
Product Type Agar Slants
Target Organism Class Fastidious Organisms
Test Type Microbial Cultivation
For Use With (Application) Isolation and cultivation of fastidious organisms, such as pathogenic Neisseria and Haemophilus from clinical and non-clinical specimens
No. per Pack 10
Packaging Type Plate
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.