Learn More
Edge Biologicals Columbia Blood Agar Plate
Catalog No. EBP3135
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
EB-P3135 10/Pk.
1 options
Catalog No. EB-P3135 Supplier Edge Biologicals Supplier No. P3135
Add to Cart
Add to Cart
General purpose medium
Columbia Blood Agar (CBA) is used as a general purpose medium for the isolation of gram-positive microorganisms from clinical specimens.
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Media Type Columbia Blood Agar
Quantity 10/Pk.