Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Columbia CNA Blood Agar Plate

Catalog No. EBP3140
Change view
Click to view available options
Certifications/Compliance:
Certificate of analysis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
EB-P3140 Certificate of analysis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EB-P3140 Supplier Edge Biologicals Supplier No. P3140
Add to Cart
Add to Cart

Selective medium

  • Color: Bright red, opaque
  • Sterile, autoclavable
  • Final pH is 7.20

Used in isolating gram-positive cocci from clinical specimens.

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Color Red
Content And Storage 8°C to 12°C
Country of Origin United States
Detects Gram-positive Cocci
Form Suspension
Format Agar Plate,
Manufacturing Origin United States
Media Type Columbia CNA
Sterility Sterile
Target Organism Class Gram-positive Cocci
Test Type Media Testing
Formulation Enzymatic digest of Casein 15g/L, Enzymatic digest of Animal tissue 8g/L, Yeast enriched Peptone 10g/L, Sodium Chloride 5g/L, Agar 15g/L, Corn Starch 1g/L, Defibrinated Sheep Blood 50mL, Nalidixic Acid 15mg/L, Colistin 10mg/L, Deionized Water 950mL
For Use With (Application) Isolation of gram positive Cocci from clinical specimens
Packaging Type Plate
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.