Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Corn Meal Agar Plate

Catalog No. EBP4145
Change view
Click to view available options
Certifications/Compliance:
Certificate of analysis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
EB-P4145 Certificate of analysis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-P4145 Supplier Edge Biologicals Supplier No. P4145
Add to Cart
Add to Cart
This item is not returnable. View return policy

General purpose medium for the cultivation of fungi

  • Prepared corn meal suspension in agar plate
  • Corn Meal infusion from solids 50g/L, Agar 15g/L, Deionized Water 1000mL
  • Color: light amber
  • Format: Suspension
  • Sterile
  • Autoclavable
  • Final pH: 5.99

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Color Light Amber
Content And Storage 8°C to 12°C
Form Liquid
Manufacturing Origin United States
Media Type Corn Meal Agar
Sterility Sterile
For Use With (Application) Cultivation of fungi
Packaging Type Plate
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.