Learn More
Edge Biologicals Prepared Plate Dehydrated Culture Media: Tyrosine Agar
Description
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Specifications
| Certifications/Compliance | Certificate of analysis |
| Content And Storage | 8°C to 12°C |
| Detects | Nocardia species |
| Form | Dehydrated |
| Format | Plate |
| Media Type | Tyrosine Agar |
| Product Type | Agar Plate |
| Formulation | Pancreatic digest of Casein 5g/L, Yeast extract 2.5g/L, Dextrose 1g/L, Agar 15g/L, Tyrosine 5g/L, Deionized Water 1000mL |
| For Use With (Application) | For differentiation of Nocardia species from clinical specimens |
| Packaging Type | Plate |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.