Learn More
Edge Biologicals Dermatophyte Test Med Plate
Description
Dermatophyte Test Medium (DTM) is used as a selective medium for the isolation of pathogenic fungi from cutaneous sources in humans and animals.
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Specifications
| Detects | Pathogenic Fungi |
| Form | Prepared Medium |
| Format | Plate |
| Media Type | Dermatophyte Test Medium |
| Product Type | Prepared Microbiology Plate |
| Target Organism Class | Fungi |
| For Use With (Application) | Isolation of pathogenic fungi |
| Packaging Type | Pack |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.