Learn More
Edge Biologicals Dermatophyte Test Medium 1oz Bottle Slant
Description
Dermatophyte Test Medium (DTM), when supplemented with Gentamicin and Chlorotetracycline, is a selective medium for the isolation of pathogenic fungi from cutaneous sources.
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Specifications
| Additive | Gentamicin, Chlorotetracycline |
| Detects | Pathogenic Fungi |
| Format | Bottle Slant |
| Media Type | Dermatophyte Test Medium |
| Product Type | Culture Medium |
| For Use With (Application) | Isolation of Pathogenic Fungi |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.