Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Heart Infusion Agar Plate

Catalog No. EBP2195
Change view
Click to view available options
Certifications/Compliance:
Certificate of analysis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
EB-P2195 Certificate of analysis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EB-P2195 Supplier Edge Biologicals Supplier No. P2195
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Beef heart infusion from 500g/L, 10g/L, Tryptose 10g/L, Sodium Chloride 5g/L, Agar 15g/L, Deionized Water 1000mL

Cultivate a wide variety of fastidious and nonfastidious microorganisms

  • General purpose medium
  • Prepared suspension in agar plate
  • Color: Clear light amber
  • Sterile
  • Autoclavable
  • pH after sterilization is 7.43

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Color Light Amber
Content And Storage 8°C to 12°C
Country of Origin United States
Detects Fastidious and non-fastidious Microorganisms
Form Liquid
Format Plate
Manufacturing Origin United States
Media Type Heart infusion Agar
Product Type Agar Plate
Sterility Sterile
Test Type Microbial Cultivation
For Use With (Application) General purpose
Packaging Type Plate
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.